Learn More
Cell Signaling Technology Matched Antibody Pair, Total Alpha-Tubulin, Human Mouse Monkey, 1 Kit
SDP
Catalog No. 502733119
Encompass
The Total α-Tubulin Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for conjugation with labels such as fluorophores, lanthanides, biotin, and beads. It is compatible with platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8))
  • BSA and azide free
  • Validated for human, mouse, and monkey species reactivity
  • Ideal for custom conjugation
  • Consists of capture and detection antibodies targeting non-overlapping epitopes
Catalog No. 50-273-3119 Supplier Cell Signaling Technology Supplier No. 54332S
The Total α-Tubulin Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for conjugation with labels such as fluorophores, lanthanides, biotin, and beads. It is compatible with platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8))
  • BSA and azide free
  • Validated for human, mouse, and monkey species reactivity
  • Ideal for custom conjugation
  • Consists of capture and detection antibodies targeting non-overlapping epitopes