Learn More
Cell Signaling Technology Total Aurora A Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503132547
Encompass
The Total Aurora A Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs suitable for custom labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, allowing for versatile conjugation with labels such as fluorophores, lanthanides, biotin, and beads.
  • Compatible with diverse platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex
  • Formulated in 1X PBS without BSA or azide
  • Validated for human species reactivity
  • Supplied as a complete kit for flexible assay development
Catalog No. 50-313-2547 Supplier Cell Signaling Technology Supplier No. 45871S
The Total Aurora A Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs suitable for custom labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, allowing for versatile conjugation with labels such as fluorophores, lanthanides, biotin, and beads.
  • Compatible with diverse platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex
  • Formulated in 1X PBS without BSA or azide
  • Validated for human species reactivity
  • Supplied as a complete kit for flexible assay development