Learn More
Cell Biolabs Inc Total Bile Acid Assay Kit (Colorimetric), 100 assays
Supplier:  Cell Biolabs Inc STA631
Encompass_Preferred
Bile acids are produced from the breakdown of cholesterol in the liver, and bile acid levels in circulation can help identify certain liver diseases. Cell Biolabs' Total Bile Acid Assay Kit is a 96-well plate-based method to measure total bile acid content in a standard 96-well colorimetric plate reader. The assay uses an enzymatic reaction in which bile acids are incubated in the presence of thio-NAD+. The kit can detect bile acid concentrations as low as 0.78 µM in serum, plasma, and cell or tissue lysates. Each kit provides sufficient reagents for 100 assays including standards and unknown samples.
Catalog No. 50-109-8796