Learn More
DIASYS DIAGNOSTIC SYSTEMS USA LLC DiaSys TruCal TBA is a lyophilized calibrator for use with Total Bile Acids assays (3x1mL)
Catalog No. NC2874340
Encompass_Preferred
DiaSys TruCal TBA is a lyophilized calibrator based on human serum/plasma. For veterinary or research use only. Contains: biological additives from bovine/ovine origin. The calibrator is used to calibrate the DiaSys Total Bile Acids 21 FS assay or any other assays from other manufacturers that are used to measure Bile Acids. DiaSys TruCal TBA must be stored refrigerated. Once reconstituted, DiaSys TruCal TBA is stable for 60 days when kept frozen.
Catalog No. NC2874340 Supplier DIASYS DIAGNOSTIC SYSTEMS USA LLC Supplier No. 111409911037
Add to Cart
Add to Cart
DiaSys TruCal TBA is a lyophilized calibrator based on human serum/plasma. For veterinary or research use only. Contains: biological additives from bovine/ovine origin. The calibrator is used to calibrate the DiaSys Total Bile Acids 21 FS assay or any other assays from other manufacturers that are used to measure Bile Acids. DiaSys TruCal TBA must be stored refrigerated. Once reconstituted, DiaSys TruCal TBA is stable for 60 days when kept frozen.