Learn More
Cell Signaling Technology Total C-Fos Matched Antibody Pair, 1 Kit
SDP
Catalog No. 503133566
Encompass
The Total c-Fos Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms. This kit provides capture and detection antibodies suitable for custom labeling, including fluorophores, lanthanides, biotin, and beads. The antibodies are provided in a BSA and azide-free formulation in 1X PBS buffer.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes
  • Compatible with platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex
  • Validated for human, mouse, and monkey reactivity
  • Supplied as a 1 kit format containing 100 μg of each antibody
  • Store at -20°C
Catalog No. 50-313-3566 Supplier Cell Signaling Technology Supplier No. 56238S
The Total c-Fos Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms. This kit provides capture and detection antibodies suitable for custom labeling, including fluorophores, lanthanides, biotin, and beads. The antibodies are provided in a BSA and azide-free formulation in 1X PBS buffer.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes
  • Compatible with platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex
  • Validated for human, mouse, and monkey reactivity
  • Supplied as a 1 kit format containing 100 μg of each antibody
  • Store at -20°C