Learn More
SPEX CERTIPREP LLC 3P Total Residual Chlorine Concentrate CRM 0.100 mg/L
Catalog No. NC0289456
Encompass_Preferred
(NSI by Zeptometrix) Convenient and easy to use. Dilute 1mL from an ampule to 1000mL to reach certified value. Always the same, lot to lot, so your dilution process remains the same minimizing errors in your lab.
Catalog No. NC0289456 Supplier SPEX CERTIPREP LLC 3P Supplier No. QCI183
 Restricted Item
Add to Cart
Add to Cart
(NSI by Zeptometrix) Convenient and easy to use. Dilute 1mL from an ampule to 1000mL to reach certified value. Always the same, lot to lot, so your dilution process remains the same minimizing errors in your lab.