Learn More
ERA WATERS CORP TOTAL CHLORINE
SDP
Catalog No. NC9016814
Encompass
One 2 mL flame-sealed ampule yields up to 2 liters after dilution. Use with titrimetric or colorimetric methods.
Catalog No. NC9016814 Supplier ERA WATERS CORP Supplier No. 501
One 2 mL flame-sealed ampule yields up to 2 liters after dilution. Use with titrimetric or colorimetric methods.