Learn More
Hach Lange™ Total Chlorine Reagent Powder Pillows
Catalog No. 501995397 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Volume (Metric):
10 mL
Catalog No. Volume (Metric)
50-199-5397 10 mL
1 options
Catalog No. 50-199-5397 Supplier Hach Lange™ Supplier No. 2105669
For total chlorine determination by the DPD method.
For use with Chlorine Visual Test Kits up to 3.5 mg/L Cl2.
Pack of 100 powder pillows. Boric Acid free!
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EPA compliant
Volume (Metric) 10 mL
For Use With (Equipment) All except DR/3000, DR/2000
Parameter Specifications Total Chlorine, Bromine, Iodine
Quantity 100 Pack
Range 0.02 to 2.0 mg/L Cl2
Test Method DPD
Type Total Chlorine Reagent Powder Pillows