Learn More
Hach Lange™ Total Chlorine Reagent Powder Pillows
Catalog No. p-8009415 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Volume (Metric):
10 mL
Catalog No. Volume (Metric)
50-199-5397 10 mL
1 options
Catalog No. 50-199-5397 Supplier Hach Lange™ Supplier No. 2105669
Only null left
Add to Cart
Add to Cart
For total chlorine determination by the DPD method.
For use with Chlorine Visual Test Kits up to 3.5 mg/L Cl2.
Pack of 100 powder pillows. Boric Acid free!
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EPA compliant
Volume (Metric) 10 mL
For Use With (Equipment) All except DR/3000, DR/2000
Parameter Specifications Total Chlorine, Bromine, Iodine
Quantity 100 Pack
Range 0.02 to 2.0 mg/L Cl2
Test Method DPD
Type Total Chlorine Reagent Powder Pillows