Learn More
HANNA INSTRUMENTS INC Total Chlorine Reagents (100 tests)
SDP
Catalog No. NC1717874
Encompass
The HI93711-01 are reagents for the colorimetric determination of total chlorine. There are enough reagents for 100 tests and are for use with Hanna's compatible benchtop and portable photometers. These high quality reagents are manufactured in our state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet for traceability.Pre-made reagents for ease of usePrepared with high purity chemicalsMarked with expiration date and lot number for traceability
Catalog No. NC1717874 Supplier HANNA INSTRUMENTS INC Supplier No. HI9371101
The HI93711-01 are reagents for the colorimetric determination of total chlorine. There are enough reagents for 100 tests and are for use with Hanna's compatible benchtop and portable photometers. These high quality reagents are manufactured in our state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet for traceability.Pre-made reagents for ease of usePrepared with high purity chemicalsMarked with expiration date and lot number for traceability