Learn More
Cayman Chemical Total ColagenHydroxyprolIn 96
SDP
Catalog No. 502893972
Encompass_Preferred
An activator of the STING pathway induces secretion of IFN-B in human PBMCs (EC50 130 nM) increases serum levels of Ifn-B Il-6 Tnf and Cxcl1 in wild-type but not Sting-/- mice at 2.5 mg/kg inhibits the cytopathic effect of HCoV-229E in infected MRC-5 cells (EC50 3 nM) decreases the level of SARS-CoV-2 RNA in primary human bronchial airway epithelial cells in an air-liquid interface assay decreases tumor volume and increases survival in a CT26 murine colorectal cancer model at 3 mg/kg this compound is expected to exist in one or both tautomeric forms
Catalog No. 50-289-3972 Supplier Cayman Chemical Supplier No. 70244096 WELL
An activator of the STING pathway induces secretion of IFN-B in human PBMCs (EC50 130 nM) increases serum levels of Ifn-B Il-6 Tnf and Cxcl1 in wild-type but not Sting-/- mice at 2.5 mg/kg inhibits the cytopathic effect of HCoV-229E in infected MRC-5 cells (EC50 3 nM) decreases the level of SARS-CoV-2 RNA in primary human bronchial airway epithelial cells in an air-liquid interface assay decreases tumor volume and increases survival in a CT26 murine colorectal cancer model at 3 mg/kg this compound is expected to exist in one or both tautomeric forms