Learn More
Alpco Ltd Total Complement Functional Screen ELISA
SDP
Supplier:  Alpco Ltd 13COMPL300
Encompass
The Total Complement Functional Screen ELISA is for the quantitative determination of functional classical, MBL and alternative complement pathways in human serum. For Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2200743