Learn More
Cell Signaling Technology Total CSF-1R/M-CSF-R Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503132335
Encompass
The Total CSF-1R/M-CSF-R Matched Antibody Pair is designed for high-throughput immunoassay platforms and applications requiring customized antibody labeling. This pair includes capture and detection antibodies that recognize non-overlapping epitopes on the CSF-1 receptor, facilitating sensitive detection in complex biological samples.
  • Compatible with diverse labeling technologies including fluorophores, lanthanides, biotin, and beads.
  • Validated for use on major platforms such as MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF).
  • Supplied in a BSA and azide-free formulation for optimal conjugation compatibility.
  • Specifically optimized for human protein detection via ELISA.
  • Consists of monoclonal antibodies to ensure high specificity and performance.
Catalog No. 50-313-2335 Supplier Cell Signaling Technology Supplier No. 32682S
The Total CSF-1R/M-CSF-R Matched Antibody Pair is designed for high-throughput immunoassay platforms and applications requiring customized antibody labeling. This pair includes capture and detection antibodies that recognize non-overlapping epitopes on the CSF-1 receptor, facilitating sensitive detection in complex biological samples.
  • Compatible with diverse labeling technologies including fluorophores, lanthanides, biotin, and beads.
  • Validated for use on major platforms such as MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF).
  • Supplied in a BSA and azide-free formulation for optimal conjugation compatibility.
  • Specifically optimized for human protein detection via ELISA.
  • Consists of monoclonal antibodies to ensure high specificity and performance.