Learn More
Cell Signaling Technology Total Cyclin E1 Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503132819
Encompass
The Total Cyclin E1 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms. This set includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for applications requiring custom antibody labeling with fluorophores, lanthanides, biotin, or beads.
  • Includes one capture antibody (100 μg) and one detection antibody (100 μg)
  • Optimized for platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex
  • Supplied in BSA and azide-free formulation
  • Validated for human species reactivity
  • Concentrated format at 1 mg/mL
Catalog No. 50-313-2819 Supplier Cell Signaling Technology Supplier No. 20905S
The Total Cyclin E1 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms. This set includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for applications requiring custom antibody labeling with fluorophores, lanthanides, biotin, or beads.
  • Includes one capture antibody (100 μg) and one detection antibody (100 μg)
  • Optimized for platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex
  • Supplied in BSA and azide-free formulation
  • Validated for human species reactivity
  • Concentrated format at 1 mg/mL