Learn More
LaMotte™ Total Dissolved and Hydrogen Sulfide Test Kit
SDPSCI_ED
Catalog No. S72404 Shop All LaMotte Company Products
Change view
Click to view available options
Range:
1ppm
Catalog No. Range
S72404 1ppm
1 options
Catalog No. S72404 Supplier LaMotte™ Supplier No. 4630
Add to Cart
Add to Cart
Titrimetric test in which total, dissolved and hydrogen sulfide can be separated.
Dropper Pipet
  • Test uses a pipet to deliver standard size drops
  • Number of drops used to obtain a color change is multiplied by a given factor to produce the test result
Specifications
For Use With (Application) Measures for total, dissolved and hydrogen sulfide in drinking water, food and beverage, industrial water boiling and cooling, laundry and sanitation, water and wastewater
No. of Tests 70
Range 1ppm
Test Method Pomeroy Methylene Blue
Type Total Dissolved and Hydrogen Sulfide Kit
WARNING: Cancer - www.P65Warnings.ca.gov