Learn More
Meso Scale Discovery Total EGFR Cytoplasmic Domain
SDP
Catalog No. 5000081720
Encompass
The Meso Scale Discovery EGFR (Epidermal Growth Factor Receptor) Assay Kit is a multiplex immunoassay that measures total EGFR protein levels in cell lysates It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample input The assay has high lot-to-lot reproducibility It can be used to monitor EGFR protein expression levels and phosphorylation states in response to treatments The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocking solution and calibrator
Catalog No. 50-000-81720 Supplier Meso Scale Discovery Supplier No. K150SPD2
The Meso Scale Discovery EGFR (Epidermal Growth Factor Receptor) Assay Kit is a multiplex immunoassay that measures total EGFR protein levels in cell lysates It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample input The assay has high lot-to-lot reproducibility It can be used to monitor EGFR protein expression levels and phosphorylation states in response to treatments The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocking solution and calibrator