Learn More
Cell Signaling Technology Total eIF2α Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503131756
Encompass
The Total eIF2α Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This pair includes capture and detection antibodies that bind to non-overlapping epitopes and are supplied free of BSA and azide, making them suitable for conjugation with labels such as fluorophores, lanthanides, biotin, and beads.
  • Ideal for platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Consists of 100 μg each of eIF2α (L57A5) mouse monoclonal antibody and eIF2α (D7D3) rabbit monoclonal antibody.
  • Validated for human species reactivity.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8).
Catalog No. 50-313-1756 Supplier Cell Signaling Technology Supplier No. 24416S
The Total eIF2α Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This pair includes capture and detection antibodies that bind to non-overlapping epitopes and are supplied free of BSA and azide, making them suitable for conjugation with labels such as fluorophores, lanthanides, biotin, and beads.
  • Ideal for platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Consists of 100 μg each of eIF2α (L57A5) mouse monoclonal antibody and eIF2α (D7D3) rabbit monoclonal antibody.
  • Validated for human species reactivity.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8).