Learn More
Perkin Elmer US LLC Total Emission Accessory
Supplier:  Perkin Elmer US LLC L2250101
Encompass_Preferred
The use of cut-off emission filters is recommended to reduce emission from other wavelengths. Five software-controlled emission filters are supplied with the LS 55 spectrometers. This accessory is compatible with LS 50/50B/55 spectrometers. It requires installation by a PerkinElmer Service Engineer.
Catalog No. 50-211-2669