Learn More
Cell Signaling Technology Total EpCAM Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503134333
Encompass
The Total EpCAM Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling. This set includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for applications needing conjugated antibodies, including platforms such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Includes 100 μg each of EpCAM (D1B3) Rabbit Monoclonal Antibody and EpCAM (VU1D9) Mouse Monoclonal Antibody
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8))
  • BSA and azide free formulation
  • Validated for human species reactivity
  • Ideal for custom labeling with fluorophores, lanthanides, biotin, or beads
Catalog No. 50-313-4333 Supplier Cell Signaling Technology Supplier No. 75612S
The Total EpCAM Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling. This set includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for applications needing conjugated antibodies, including platforms such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Includes 100 μg each of EpCAM (D1B3) Rabbit Monoclonal Antibody and EpCAM (VU1D9) Mouse Monoclonal Antibody
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8))
  • BSA and azide free formulation
  • Validated for human species reactivity
  • Ideal for custom labeling with fluorophores, lanthanides, biotin, or beads