Learn More
Air Science Usa Total Exhaust Fume Hood 24" Wide, Polypropylene
SDP
Catalog No. NC3835596
Encompass_Preferred
PurairTotal Exhaust Series Fume Hood, 24" / 600mm Width, Polypropylene Construction, 115V 60Hz, North American Cord Set (unless specified)
Catalog No. NC3835596 Supplier Air Science Usa Supplier No. PTEFH24A
PurairTotal Exhaust Series Fume Hood, 24" / 600mm Width, Polypropylene Construction, 115V 60Hz, North American Cord Set (unless specified)