Learn More
Innovative Research Inc Human Total Factor VIII ELISA Kit
Supplier:  Innovative Research Inc IHUFVIIIKTT
Encompass_Preferred
This human coagulation Factor VIII antigen assay is intended for the quantitative determination of total Factor VIII antigen in human plasma. For research use only. Strip well format. Reagents for up to 96 tests.
Catalog No. 50-203-6110