Learn More
Cell Signaling Technology Total FGFR2 Matched Antibody Pair, Human, Mouse, 1 Kit
SDP
Catalog No. 503131618
Encompass
The Total FGF Receptor 2 Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling, including biotin, fluorophores, lanthanides, and beads. Compatible platforms include MSD, Quanterix Simoa, Luminex, and Alpha Technology such as AlphaScreen, AlphaLISA, LANCE, and HTRF.
  • Supplied in BSA and azide-free formulation for optimal labeling.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes.
  • Validated for reactivity in human and mouse samples.
  • Suitable for applications including ELISA.
  • Formulated in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl) at pH 7.8.
Catalog No. 50-313-1618 Supplier Cell Signaling Technology Supplier No. 99057S
The Total FGF Receptor 2 Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling, including biotin, fluorophores, lanthanides, and beads. Compatible platforms include MSD, Quanterix Simoa, Luminex, and Alpha Technology such as AlphaScreen, AlphaLISA, LANCE, and HTRF.
  • Supplied in BSA and azide-free formulation for optimal labeling.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes.
  • Validated for reactivity in human and mouse samples.
  • Suitable for applications including ELISA.
  • Formulated in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl) at pH 7.8.