Learn More
Cell Signaling Technology Matched Antibody Pair, Total FGFR4, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503133680
Encompass
This kit provides a total FGF receptor 4 matched antibody pair designed for use in immunoassays and high-throughput ELISA platforms, including MSD, Quanterix Simoa, Alpha Technology, and Luminex. The system includes capture and detection antibodies that bind to non-overlapping epitopes, allowing for versatile custom labeling with fluorophores, lanthanides, biotin, or beads.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8))
  • BSA and azide free formulation
  • Optimized for ELISA applications
  • Human reactivity
  • Includes 100 μg each of rabbit and mouse monoclonal antibodies
  • Store at -20°C
Catalog No. 50-313-3680 Supplier Cell Signaling Technology Supplier No. 47216S
This kit provides a total FGF receptor 4 matched antibody pair designed for use in immunoassays and high-throughput ELISA platforms, including MSD, Quanterix Simoa, Alpha Technology, and Luminex. The system includes capture and detection antibodies that bind to non-overlapping epitopes, allowing for versatile custom labeling with fluorophores, lanthanides, biotin, or beads.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8))
  • BSA and azide free formulation
  • Optimized for ELISA applications
  • Human reactivity
  • Includes 100 μg each of rabbit and mouse monoclonal antibodies
  • Store at -20°C