Learn More
Cell Signaling Technology Total FLT3 Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503131815
Encompass
The Total FLT3 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs suitable for specialized or custom labeling. This pair includes capture and detection antibodies that bind to non-overlapping epitopes and are supplied in a BSA and Azide-free formulation, making them compatible with various conjugation methods including fluorophores, lanthanides, biotin, and beads.
  • Compatible with platforms such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex
  • Includes capture and detection antibodies that target non-overlapping epitopes
  • Supplied in a BSA and Azide-free formulation for flexible downstream labeling
  • Validated for human species reactivity
  • Ideal for applications requiring custom or specialized antibody conjugation
Catalog No. 50-313-1815 Supplier Cell Signaling Technology Supplier No. 20510S
The Total FLT3 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs suitable for specialized or custom labeling. This pair includes capture and detection antibodies that bind to non-overlapping epitopes and are supplied in a BSA and Azide-free formulation, making them compatible with various conjugation methods including fluorophores, lanthanides, biotin, and beads.
  • Compatible with platforms such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex
  • Includes capture and detection antibodies that target non-overlapping epitopes
  • Supplied in a BSA and Azide-free formulation for flexible downstream labeling
  • Validated for human species reactivity
  • Ideal for applications requiring custom or specialized antibody conjugation