Learn More
Cell Signaling Technology Total FoxO1 Matched Antibody Pair, Unconjugated, Human and Mouse, 1 Kit
SDP
Catalog No. 503132149
Encompass
This Total FoxO1 Matched Antibody Pair is designed for use with high-throughput immunoassay platforms that require custom antibody labeling. These pairs utilize antibodies that bind to non-overlapping epitopes and are supplied in a BSA- and azide-free formulation, making them suitable for conjugation with fluorophores, lanthanides, biotin, or beads.
  • Compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied as 100 μg each of rabbit and mouse monoclonal antibodies.
  • Formulated in 1X PBS (pH 7.8).
  • Validated for reactivity in human and mouse species.
  • Optimized for ELISA applications.
  • Requires storage at -20°C.
Catalog No. 50-313-2149 Supplier Cell Signaling Technology Supplier No. 30452S
This Total FoxO1 Matched Antibody Pair is designed for use with high-throughput immunoassay platforms that require custom antibody labeling. These pairs utilize antibodies that bind to non-overlapping epitopes and are supplied in a BSA- and azide-free formulation, making them suitable for conjugation with fluorophores, lanthanides, biotin, or beads.
  • Compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied as 100 μg each of rabbit and mouse monoclonal antibodies.
  • Formulated in 1X PBS (pH 7.8).
  • Validated for reactivity in human and mouse species.
  • Optimized for ELISA applications.
  • Requires storage at -20°C.