Learn More
Hach Company Free and Total Chlorine TNTplus Vial Test (0.05-2.00 mg/L Cl₂), 24 Tests
Catalog No. 501998152 Shop All Hach Company Products
Encompass
Free and Total Chlorine TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Free Chlorine by the DPD Method.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-8152 Supplier Hach Company Supplier No. TNT867
Free and Total Chlorine TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Free Chlorine by the DPD Method.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.