Learn More
Hach Company Titration Application Pack Water: Total Hardness (Color)
Supplier:  Hach Company AP0004.AT1112
Encompass
Application Pack for the determination of Calcium and Magnesium Hardness in water. Suited for TitraLab AT1000 Series, Automatic Titrator Models AT1112/AT1122/AT1222.
Catalog No. 50-199-7509