Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Total Hardness Indicator Eriochrome Black T Indicator, Dry Mix, Reagents

Catalog No. 01335317
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-317 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-317 Supplier Reagents Holdings Llc Supplier No. CE255600100C2
 Restricted Item
Add to Cart
Add to Cart

Total Hardness Indicator Eriochrome Black T Indicator, Dry Mix, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Total Hardness Indicator Eriochrome Black T Indicator, Dry Mix
CAS 7778-80-5, 7778-77-0
Grade Reagent
Packaging Glass Bottle
Specific Gravity 2.372
Color Purple
Quantity 100 g
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.