Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Total Hardness Indicator, Ricca Chemical
SDP

Catalog No. 8669100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
White Poly Wide Mouth Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8669100 100 g White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 8669100 Supplier Ricca Chemical Company Supplier No. 8669100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Total Hardness Indicator
CAS 7778-80-5, 5470-11-1, 7778-77-0, 1787-61-7
CAS Min % 77.0, 16.66, 2.94, 0.98
CAS Max % 81.0, 17.34, 3.06, 1.02
Packaging White Poly Wide Mouth Bottle
Molecular Formula ClH4NO
MDL Number MFCD00051089
Color Red
Quantity 100 g
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
SMILES Cl.NO
Physical Form Powder
IUPAC Name hydroxylamine hydrochloride
Molecular Weight (g/mol) Mixture
Grade Indicator
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.