Learn More
LaMotte™ Total Hardness Test Kit
SDPSCI_ED
Catalog No. S72403 Shop All LaMotte Company Products
Change view
Click to view available options
Range:
10ppm
Catalog No. Range
S72403 10ppm
1 options
Catalog No. S72403 Supplier LaMotte™ Supplier No. 4482LT02
Add to Cart
Add to Cart
Uses EDTA titration
Dropper Bottles - Bottle tips deliver a consistent standard drop size to add titrant to the sample; the number of drops used to complete the reaction is multiplied by a given equivalence factor to determine the concentration.
  • Hardness - Test uses EDTA titration with a red to blue endpoint; with inhibitors to eliminate metal interferences. Liquid buffer and tablet indicator. Results as CaCO3.
Specifications
For Use With (Application) Measures for total hardness in food and beverage, industrial water boiling and cooling, laundry, sanitation, water and wastewater
No. of Tests 50
Range 10ppm
Test Method EDTA Titration
Type Total Hardness Test Kit