Learn More
Cell Signaling Technology Matched Antibody Pair, Total HER2/ErbB2, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503134197
Encompass
The Total HER2/ErbB2 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for applications such as MSD, Quanterix Simoa, Alpha Technology, and Luminex. The antibodies are provided in a BSA and azide-free formulation, allowing for versatile conjugation with fluorophores, lanthanides, biotin, or beads.
  • Validated for ELISA applications
  • Consists of capture and detection antibodies binding to non-overlapping epitopes
  • Provided in a 1X PBS (pH 7.8) formulation
  • BSA and azide-free
  • Compatible with diverse conjugation methods
Catalog No. 50-313-4197 Supplier Cell Signaling Technology Supplier No. 56895S
The Total HER2/ErbB2 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for applications such as MSD, Quanterix Simoa, Alpha Technology, and Luminex. The antibodies are provided in a BSA and azide-free formulation, allowing for versatile conjugation with fluorophores, lanthanides, biotin, or beads.
  • Validated for ELISA applications
  • Consists of capture and detection antibodies binding to non-overlapping epitopes
  • Provided in a 1X PBS (pH 7.8) formulation
  • BSA and azide-free
  • Compatible with diverse conjugation methods