Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Total Insulin Receptor Beta Matched Antibody Pair, Unconjugated, Human, 1 Kit
SDP

Catalog No. 503131748
Encompass

The Total Insulin Receptor β Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling, including fluorophores, lanthanides, biotin, and beads. The capture and detection antibodies in this kit bind to non-overlapping epitopes and are compatible with platforms such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.

  • Ideal for applications requiring custom antibody conjugation
  • Formulated in 1X PBS, BSA and azide free
  • Species reactivity is limited to human
  • Storage recommended at -20°C in single-use aliquots to avoid freeze/thaw cycles
  • Includes 100 μg each of insulin receptor β (L55B10) mouse monoclonal antibody and insulin receptor β (A6H3G) rabbit monoclonal antibody

Catalog No. 50-313-1748 Supplier Cell Signaling Technology Supplier No. 81512S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Total Insulin Receptor β Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling, including fluorophores, lanthanides, biotin, and beads. The capture and detection antibodies in this kit bind to non-overlapping epitopes and are compatible with platforms such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.

  • Ideal for applications requiring custom antibody conjugation
  • Formulated in 1X PBS, BSA and azide free
  • Species reactivity is limited to human
  • Storage recommended at -20°C in single-use aliquots to avoid freeze/thaw cycles
  • Includes 100 μg each of insulin receptor β (L55B10) mouse monoclonal antibody and insulin receptor β (A6H3G) rabbit monoclonal antibody

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.