Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TISAB Concentrate, Fisherbrand
SDP

Catalog No. ACCU0831500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACCU0831500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ACCU0831500
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material Total Ionic Strength Adjustment Buffer (TISAB) Concentrate
Packaging Natural Poly Bottle
CAS 1310-73-2, 13291-61-7, 64-19-7, 7732-18-5
Odor Odorless to slight acetic acid odor
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
Solubility Miscible with water
Solution Type Calibration Solution
Quantity 500 mL
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.