Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Total Ionic Strength Adjustment Buffer (TISAB II), with CDTA, Fisherbrand
SDP

Catalog No. ACCU0835500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACCU0835500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ACCU0835500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for Fluoride Analysis using Ion Selective Electrodes

Specifications

Chemical Name or Material Total Ionic Strength Adjustment Buffer (TISAB)
Packaging Natural Poly Bottle
CAS 1310-73-2, 13291-61-7, 64-19-7, 7647-14-5, 7732-18-5
Odor Slight acetic acid odor
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
Solubility Miscible with water
Solution Type Calibration Solution
Quantity 500 mL
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.