Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Total Ionic Strength Adjustment Buffer (TISAB IV), with Tartrate, Ricca Chemical
SDP

Catalog No. 867332 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8673-32 1 L
8673-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 8673-32 Supplier Ricca Chemical Company Supplier No. 867332
Add to Cart
Add to Cart

For Analysis of Fluoride Analysis Using Ion Selective Electrodes

Specifications

Chemical Name or Material Buffer Solution, Total Ionic Strength Adjustment Buffer (TISAB IV)
Name Note With Tartrate
CAS 7732-18-5, 77-86-1, 6106-24-7, 7647-01-0
CAS Min % 62.0, 15.1, 14.35, 6.18
CAS Max % 65.0, 15.72, 14.94, 6.43
Color Yellow to Green
Concentration 1X
For Use With (Application) Fluoride Analysis
For Use With (Equipment) pH Meter and Electrode
pH 9
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Natural Poly Bottle
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.