Learn More
Cell Signaling Technology Total IRF-3 Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503131594
Encompass
The Total IRF-3 Matched Antibody Pair is designed for high-throughput immunoassays and ELISA platforms that require specialized labeling. The pair contains capture and detection antibodies that bind to non-overlapping epitopes, allowing for versatile conjugation with fluorophores, lanthanides, biotin, or beads.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl) at pH 7.8
  • Formulated BSA and azide free
  • Validated for human reactivity
  • Designed for ELISA applications
  • Consists of 100 μg each of rabbit monoclonal capture and mouse monoclonal detection antibodies
Catalog No. 50-313-1594 Supplier Cell Signaling Technology Supplier No. 58254S
The Total IRF-3 Matched Antibody Pair is designed for high-throughput immunoassays and ELISA platforms that require specialized labeling. The pair contains capture and detection antibodies that bind to non-overlapping epitopes, allowing for versatile conjugation with fluorophores, lanthanides, biotin, or beads.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl) at pH 7.8
  • Formulated BSA and azide free
  • Validated for human reactivity
  • Designed for ELISA applications
  • Consists of 100 μg each of rabbit monoclonal capture and mouse monoclonal detection antibodies