Learn More
Cayman Chemical Total Iron-BIndng CapacIty 1ea
SDP
Catalog No. 502894035
Encompass_Preferred
Cayman’s Total Iron-Binding Capacity (TIBC) and Serum Iron Assay Kit provides a convenient colorimetric method of determining iron levels and UIBC in serum. Measurement of both serum iron and UIBC allows for the calculation of TIBC, which is an estimation of transferrin levels in the serum. This assay has a range of 0-100 µM (0-560 UG/dl) and a lower limit of detection (LLOD) of 0.68 µM (3.8 UG/dl).
Catalog No. 50-289-4035 Supplier Cayman Chemical Supplier No. 7022301 EA
Cayman’s Total Iron-Binding Capacity (TIBC) and Serum Iron Assay Kit provides a convenient colorimetric method of determining iron levels and UIBC in serum. Measurement of both serum iron and UIBC allows for the calculation of TIBC, which is an estimation of transferrin levels in the serum. This assay has a range of 0-100 µM (0-560 UG/dl) and a lower limit of detection (LLOD) of 0.68 µM (3.8 UG/dl).