Learn More
Cell Signaling Technology Total LRRK2 Matched Antibody Pair, Human, Mouse, Unconjugated, 1 Kit
SDP
Catalog No. 503131898
Encompass
The Total LRRK2 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms, including MSD, Quanterix Simoa, Alpha Technology, and Luminex. This kit is suitable for applications requiring custom antibody labeling with fluorophores, lanthanides, biotin, or beads.
  • Includes 100 μg each of LRRK2 (D18E12) rabbit monoclonal antibody and LRRK2 (E7P4V) mouse monoclonal antibody
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8)
  • BSA and azide free formulation
  • Compatible with human and mouse samples
  • Optimized for ELISA applications
  • Consists of capture and detection antibodies binding to non-overlapping epitopes
Catalog No. 50-313-1898 Supplier Cell Signaling Technology Supplier No. 65040S
The Total LRRK2 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms, including MSD, Quanterix Simoa, Alpha Technology, and Luminex. This kit is suitable for applications requiring custom antibody labeling with fluorophores, lanthanides, biotin, or beads.
  • Includes 100 μg each of LRRK2 (D18E12) rabbit monoclonal antibody and LRRK2 (E7P4V) mouse monoclonal antibody
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8)
  • BSA and azide free formulation
  • Compatible with human and mouse samples
  • Optimized for ELISA applications
  • Consists of capture and detection antibodies binding to non-overlapping epitopes