Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Total Met Matched Antibody Pair, Human, 1 Kit
SDP

Catalog No. 503134318
Encompass

The Total Met Matched Antibody Pair provides a optimized set of capture and detection antibodies designed for use in immunoassay technologies, such as ELISA platforms. These antibodies recognize non-overlapping epitopes on the Met protein, facilitating precise quantification. The pair is supplied in a BSA and azide-free formulation, making it suitable for custom conjugation with various labels, including biotin, fluorophores, and beads.

  • Validated for use in ELISA applications
  • Contains capture and detection antibodies in a BSA and azide-free formulation
  • Compatible with high-throughput platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex
  • Species reactivity is validated for human
  • Supplied in 1X PBS (10 mM Na&sub2;HPO&sub4;, 3 mM KCl, 2 mM KH&sub2;PO&sub4;, and 140 mM NaCl (pH 7.8))

Catalog No. 50-313-4318 Supplier Cell Signaling Technology Supplier No. 23910S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Total Met Matched Antibody Pair provides a optimized set of capture and detection antibodies designed for use in immunoassay technologies, such as ELISA platforms. These antibodies recognize non-overlapping epitopes on the Met protein, facilitating precise quantification. The pair is supplied in a BSA and azide-free formulation, making it suitable for custom conjugation with various labels, including biotin, fluorophores, and beads.

  • Validated for use in ELISA applications
  • Contains capture and detection antibodies in a BSA and azide-free formulation
  • Compatible with high-throughput platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex
  • Species reactivity is validated for human
  • Supplied in 1X PBS (10 mM Na&sub2;HPO&sub4;, 3 mM KCl, 2 mM KH&sub2;PO&sub4;, and 140 mM NaCl (pH 7.8))

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.