Learn More
Grainger Lovibond Photometer Reagent Set, Total Nitrogen, 10-150 mg/L, 25 Tests
Catalog No. 502757820
Encompass_Preferred
This Total Nitrogen HR Reagent Set is designed for the accurate measurement of total nitrogen in water samples. It is intended for use with colorimeters and photometers to determine nitrogen levels within a range of 10 to 150 mg/L N.
  • Measurement range of 10 to 150 mg/L N
  • Includes necessary vials and reagents for water analysis
  • Compatible with photometers and colorimeters
  • Lab grade chemical reagents
  • Maximum storage temperature of 20°C
  • Hazard classification: Met. Corr. 1
Catalog No. 50-275-7820 Supplier Grainger Supplier No. 15F957
This Total Nitrogen HR Reagent Set is designed for the accurate measurement of total nitrogen in water samples. It is intended for use with colorimeters and photometers to determine nitrogen levels within a range of 10 to 150 mg/L N.
  • Measurement range of 10 to 150 mg/L N
  • Includes necessary vials and reagents for water analysis
  • Compatible with photometers and colorimeters
  • Lab grade chemical reagents
  • Maximum storage temperature of 20°C
  • Hazard classification: Met. Corr. 1