Learn More
Aqua Solutions Total Organic Carbon Std. 10,000 ppm, Preserved with H₂SO₄(250ML)
SDP
Catalog No. NC9670175
Encompass_Preferred
Total Organic Carbon Std. 10,000 ppm, Preserved with H₂SO₄(250ML)
Catalog No. NC9670175 Supplier Aqua Solutions Supplier No. DC804130
Total Organic Carbon Std. 10,000 ppm, Preserved with H₂SO₄(250ML)