Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Total Petroleum Hydrocarbons (TPH) Mixture 3 1,000 μg/mL each component in carbon disulfide, MilliporeSigma™ Supelco™

Catalog No. 111016514 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6514 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6514 Supplier MilliporeSigma Supelco Supplier No. 861394U

missing translation for 'validMainframeMsg'

Petroleum
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 75-15-0
Flash Point -30°C - Closed Cup
Type Analytical Standard
Quantity 1 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 1,000 μg/mL each component in carbon disulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.