Learn More
Innovative Research Inc Mouse Total Plasminogen ELISA Kit
Supplier:  Innovative Research Inc IMSPLGKTT
Encompass_Preferred
This mouse plasminogen total assay is for the quantitative determination of total plasminogen and plasmin in mouse plasma, serum, urine, cell culture media, or tissue extracts. For research use only. Strip well format. Reagents for up to 96 tests.
Catalog No. 50-203-6181
 Restricted Item