Learn More
Crystal Chem Inc Total Proinsulin Chemiluminescence Kit, 5 Kit Pack, 96-wells per kit
SDP
Catalog No. 501947927
Encompass
The Total Proinsulin Chemiluminescence is a highly sensitive assay that is used to quantify levels of human total proinsulin in serum and plasma. Highlights: Kits use a sample volume of only 50 µL. Highly specific for total proinsulin. Dynamic range of 0.25 - 220 pmol/mL. Highly precise with low CV's. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7927 Supplier Crystal Chem Inc Supplier No. 90112
The Total Proinsulin Chemiluminescence is a highly sensitive assay that is used to quantify levels of human total proinsulin in serum and plasma. Highlights: Kits use a sample volume of only 50 µL. Highly specific for total proinsulin. Dynamic range of 0.25 - 220 pmol/mL. Highly precise with low CV's. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.