Learn More
Verichem Laboratories TOTAL PROTEIN/ALBUMIN STD KIT
Catalog No. NC9592198
Encompass_Preferred
Total Protein / albumin Standard Kit. 5x15 mL aEc for calibration verification or calibration aEc prepared with human serum albumin aEc accurate, liquid standards containing purified proteins in saline aEc stable 24 months from date of manufacture aEc known concentrations verified against available NIST Standard Reference Material
Catalog No. NC9592198 Supplier Verichem Laboratories Supplier No. 9460
Total Protein / albumin Standard Kit. 5x15 mL aEc for calibration verification or calibration aEc prepared with human serum albumin aEc accurate, liquid standards containing purified proteins in saline aEc stable 24 months from date of manufacture aEc known concentrations verified against available NIST Standard Reference Material