Learn More
MilliporeSigma TOTAL PROTEIN EXTRACTION KIT
Catalog No. 50814091
Encompass
Compartmental Protein Extraction Kit, For extracting cytoplasmic, Nuclear, Membrane and cytoskeletal proteins from tissues and cells, Contains Buffer C, W, N, M and CS, Store Buffers at 2 deg. to 8 deg.C for up to 12 months, 1 KT
Catalog No. 50-814-091 Supplier MilliporeSigma Supplier No. 2145
Compartmental Protein Extraction Kit, For extracting cytoplasmic, Nuclear, Membrane and cytoskeletal proteins from tissues and cells, Contains Buffer C, W, N, M and CS, Store Buffers at 2 deg. to 8 deg.C for up to 12 months, 1 KT