Learn More
Sigma Aldrich Fine Chemicals Biosciences Total Protein Kit Micro Lo
Catalog No. 502904150
Encompass
This method utilizes sodium dodecylsulfate included in the Lowry reagent to facilitate the disSOLUTIONon of relatively insoluble lipoproteins. An optional deoxycholate-TCA precipitation step permits removal of interfering substances such as Tris sucrose EDTA salts etc.
Catalog No. 50-290-4150 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. TP03001KT
This method utilizes sodium dodecylsulfate included in the Lowry reagent to facilitate the disSOLUTIONon of relatively insoluble lipoproteins. An optional deoxycholate-TCA precipitation step permits removal of interfering substances such as Tris sucrose EDTA salts etc.