Learn More
Biochain Institute Inc Total Protein - Multiple Sclerosis Disease: Brain: Cerebellum, 1 mg/PK
SDP
Catalog No. 50183520
Encompass_Preferred
Tissue total protein is prepared from whole tissue homogenates. It presents a consistent pattern on SDS-PAGE analysis. The total protein is stored in a buffer with protease inhibitor cocktail, and provided at a concentration of 5 mg/ml.
Catalog No. 50-183-520 Supplier Biochain Institute Inc Supplier No. P1236039MSC
Add to Cart
Add to Cart
Tissue total protein is prepared from whole tissue homogenates. It presents a consistent pattern on SDS-PAGE analysis. The total protein is stored in a buffer with protease inhibitor cocktail, and provided at a concentration of 5 mg/ml.