Learn More
Biochain Institute Inc Total Protein - Mouse Normal Tissue: Skeletal Muscle, 1 mg/PK
SDP
Catalog No. 50183770
Encompass_Preferred
Tissue total protein is prepared from whole tissue homogenates. It presents a consistent pattern on SDS-PAGE analysis. The total protein is stored in a buffer with protease inhibitor cocktail, and provided at a concentration of 5 mg/ml.
Catalog No. 50-183-770 Supplier Biochain Institute Inc Supplier No. P1334171
Tissue total protein is prepared from whole tissue homogenates. It presents a consistent pattern on SDS-PAGE analysis. The total protein is stored in a buffer with protease inhibitor cocktail, and provided at a concentration of 5 mg/ml.