Learn More
ProteinSimple Total Protein Streptavidin-HRP
Supplier:  ProteinSimple 042976
Encompass
Use with Compass v2.6 or higher. Streptavidin-HRP for Total Protein Assay compatible with Jess, Abby, Wes, Peggy Sue and Sally Sue systems., 2.0 mL.
Catalog No. NC1072362